siRNA Details
| Gene Symbol: Upp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106481.1 | NP_001099951.1 | 4 | 835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upp2| Gene Symbol: Upp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106481.1 | NP_001099951.1 | 4 | 835 | ||