siRNA Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013916.1 | NP_001013938.1 | 2 | 342 | ||
| GenBank mRNA | BC089829.1 | - | 2 | 342 | 7322853 | |
| FQ209402.1 | - | 2 | 118 | |||
| FQ213169.1 | - | 2 | 136 | |||
| FQ216383.1 | - | 2 | 124 | |||
| FQ219212.1 | - | 2 | 149 | |||
| FQ219294.1 | - | 2 | 122 | |||
| FQ219651.1 | - | 2 | 122 | |||
| FQ219714.1 | - | 2 | 130 | |||

Cd302
Genomic Map