siRNA Details
| Gene Symbol: Spc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009654.1 | NP_001009654.1 | 4, 5 | 430 | ||
| XM_006234288.2 | XP_006234350.1 | 4, 5 | 436 | |||
| GenBank mRNA | BC088213.1 | - | 4, 5 | 430 | 7383479 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spc25| Gene Symbol: Spc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009654.1 | NP_001009654.1 | 4, 5 | 430 | ||
| XM_006234288.2 | XP_006234350.1 | 4, 5 | 436 | |||
| GenBank mRNA | BC088213.1 | - | 4, 5 | 430 | 7383479 | |