siRNA Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106485.3 | NP_001099955.2 | 3 | 508 | ||
| XM_006234400.2 | XP_006234462.1 | 3 | 711 | |||
| GenBank mRNA | FQ229205.1 | - | 3 | 492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp7| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106485.3 | NP_001099955.2 | 3 | 508 | ||
| XM_006234400.2 | XP_006234462.1 | 3 | 711 | |||
| GenBank mRNA | FQ229205.1 | - | 3 | 492 | ||