siRNA Details
| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106486.2 | NP_001099956.2 | 3, 4 | 629 | ||
| GenBank mRNA | CA338978.1 | - | 2, 3 | 235 | ||
| DY315949.1 | - | 3, 4 | 562 | |||
| FQ226920.1 | - | 2, 3 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc10| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106486.2 | NP_001099956.2 | 3, 4 | 629 | ||
| GenBank mRNA | CA338978.1 | - | 2, 3 | 235 | ||
| DY315949.1 | - | 3, 4 | 562 | |||
| FQ226920.1 | - | 2, 3 | 261 | |||