siRNA Details
| Gene Symbol: Frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100527.1 | NP_001093997.1 | 6 | 1227 | ||
| XM_006234429.2 | XP_006234491.1 | 5 | 1651 | |||
| GenBank mRNA | BC098777.1 | - | 3 | 325 | ||
| BC166428.1 | - | 6 | 1227 | 7106784 | ||
More Information
Other Species Targeted
Set Membership: |
|

Frzb
Genomic Map