siRNA Details
| Gene Symbol: Serping1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199093.1 | NP_954524.1 | 2 | 268 | ||
| XM_006234447.2 | XP_006234509.1 | 3 | 343 | |||
| GenBank mRNA | BC061860.1 | - | 2 | 268 | 5598525 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serping1| Gene Symbol: Serping1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199093.1 | NP_954524.1 | 2 | 268 | ||
| XM_006234447.2 | XP_006234509.1 | 3 | 343 | |||
| GenBank mRNA | BC061860.1 | - | 2 | 268 | 5598525 | |