siRNA Details
| Gene Symbol: Serping1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199093.1 | NP_954524.1 | 4 | 839 | ||
| XM_006234447.2 | XP_006234509.1 | 5 | 914 | |||
| GenBank mRNA | BC061860.1 | - | 4 | 839 | 5598525 | |
| FQ219227.1 | - | 3 | 543 | |||

Serping1
Genomic Map