siRNA Details
| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106487.1 | NP_001099957.1 | 2 | 437 | ||
| XM_006234449.1 | XP_006234511.1 | 3 | 473 | |||
| XM_006234450.1 | XP_006234512.1 | 3 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prg3| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106487.1 | NP_001099957.1 | 2 | 437 | ||
| XM_006234449.1 | XP_006234511.1 | 3 | 473 | |||
| XM_006234450.1 | XP_006234512.1 | 3 | 456 | |||