siRNA Details
| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234484.2 | XP_006234546.1 | 6 | 3902 | ||
| XM_006234485.2 | XP_006234547.1 | 6 | 4382 | |||
| XM_006234486.2 | XP_006234548.1 | 6 | 3845 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnks1bp1| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234484.2 | XP_006234546.1 | 6 | 3902 | ||
| XM_006234485.2 | XP_006234547.1 | 6 | 4382 | |||
| XM_006234486.2 | XP_006234548.1 | 6 | 3845 | |||