siRNA Details
| Gene Symbol: Olr466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000298.1 | NP_001000298.1 | 1 | 251 | ||
| Gene Symbol: Olr465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000297.1 | NP_001000297.1 | 1 | 251 | ||

Olr466
Genomic Map