siRNA Details
Gene Symbol: Olr470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000300.1 | NP_001000300.1 | 1 | 256 |
Gene Symbol: LOC684683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749504.2 | XP_003749552.1 | 1 | 311 |
