siRNA Details
| Gene Symbol: Olr515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000315.1 | NP_001000315.1 | 1 | 245 | ||
| Gene Symbol: Olr519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000318.1 | NP_001000318.1 | 1 | 251 | ||
More Information
Set Membership: |
|

Olr515
Genomic Map