siRNA Details
| Gene Symbol: Olr515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000315.1 | NP_001000315.1 | 1 | 159 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr515| Gene Symbol: Olr515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000315.1 | NP_001000315.1 | 1 | 159 | ||
Set Membership: | 
		   
					
						
							 |