siRNA Details
| Gene Symbol: Olr528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000319.1 | NP_001000319.1 | 1 | 499 | ||
| Gene Symbol: Olr522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000562.1 | NP_001000562.1 | 1 | 499 | ||

Olr528
Genomic Map