siRNA Details
| Gene Symbol: Olr576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000328.1 | NP_001000328.1 | 1 | 387 | ||
| Gene Symbol: Olr567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000326.1 | NP_001000326.1 | 1 | 387 | ||

Olr576
Genomic Map