siRNA Details
| Gene Symbol: Olr597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000331.1 | NP_001000331.1 | 419 | |||
| Gene Symbol: LOC100911348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749512.3 | XP_003749560.2 | 1 | 419 | ||

Olr597
Genomic Map