siRNA Details
| Gene Symbol: Olr631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000339.1 | NP_001000339.1 | 1 | 791 | ||
| Gene Symbol: Olr623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000651.1 | NP_001000651.1 | 1 | 791 | ||
| Gene Symbol: Olr625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000649.1 | NP_001000649.1 | 1 | 791 | ||

Olr631
Genomic Map