siRNA Details
| Gene Symbol: Olr651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000341.1 | NP_001000341.1 | 1 | 530 | ||
| Gene Symbol: Olr650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000639.1 | NP_001000639.1 | 1 | 530 | ||

Olr651
Genomic Map