siRNA Details
| Gene Symbol: Olr657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000343.1 | NP_001000343.1 | 1 | 721 | ||
| Gene Symbol: Olr662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001060.1 | NP_001001060.1 | 1 | 721 | ||

Olr657
Genomic Map