siRNA Details
| Gene Symbol: Olr658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000344.1 | NP_001000344.1 | 1 | 637 | ||
| Gene Symbol: Olr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234496.2 | XP_006234558.2 | 1 | 637 | ||

Olr658
Genomic Map