siRNA Details
| Gene Symbol: Olr673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000351.1 | NP_001000351.1 | 427 | |||
| Gene Symbol: Olr671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001061.1 | NP_001001061.1 | 1 | 427 | ||

Olr673
Genomic Map