siRNA Details
| Gene Symbol: Olr674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000352.1 | NP_001000352.1 | 1 | 625 | ||
| Gene Symbol: Olr672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000350.1 | NP_001000350.1 | 1 | 625 | ||
| Gene Symbol: Olr675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000632.1 | NP_001000632.1 | 1 | 625 | ||

Olr674
Genomic Map