siRNA Details
| Gene Symbol: Olr679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000354.1 | NP_001000354.1 | 1 | 331 | ||
| Gene Symbol: Olr681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000631.1 | NP_001000631.1 | 1 | 331 | ||

Olr679
Genomic Map