siRNA Details
| Gene Symbol: Olr684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001378.1 | NP_001001378.1 | 1 | 637 | ||
| Gene Symbol: Olr705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000570.1 | NP_001000570.1 | 1 | 637 | ||

Olr684
Genomic Map