siRNA Details
| Gene Symbol: Olr728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000363.1 | NP_001000363.1 | 1 | 802 | ||
| Gene Symbol: Olr726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000620.2 | NP_001000620.2 | 2 | 745 | ||

Olr728
Genomic Map