siRNA Details
| Gene Symbol: Mtch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106488.2 | NP_001099958.2 | 9 | 633 | ||
| GenBank mRNA | BC158687.1 | - | 9 | 584 | 8375010 | |
| CK366546.1 | - | 30 | ||||
| DY311538.1 | - | 584 | ||||
| EV779017.1 | - | 654 | ||||

Mtch2
Genomic Map