siRNA Details
| Gene Symbol: RGD1309540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013918.1 | NP_001013940.1 | 170 | |||
| GenBank mRNA | BC087159.1 | - | 170 | 7134306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309540| Gene Symbol: RGD1309540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013918.1 | NP_001013940.1 | 170 | |||
| GenBank mRNA | BC087159.1 | - | 170 | 7134306 | ||