siRNA Details
| Gene Symbol: Apip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106492.1 | NP_001099962.1 | 5, 6 | 579 | ||
| XM_006234638.2 | XP_006234700.1 | 5, 6 | 628 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apip| Gene Symbol: Apip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106492.1 | NP_001099962.1 | 5, 6 | 579 | ||
| XM_006234638.2 | XP_006234700.1 | 5, 6 | 628 | |||