siRNA Details
| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024981.1 | NP_001020152.1 | 5 | 718 | ||
| XM_006234686.2 | XP_006234748.1 | 6 | 890 | |||
| XM_006234687.2 | XP_006234749.1 | 5 | 789 | |||
| XM_006234688.2 | XP_006234750.1 | 6 | 885 | |||
| XM_006234689.2 | XP_006234751.1 | 6 | 864 | |||
| XM_008762084.1 | XP_008760306.1 | 6 | 973 | |||
| XM_008762085.1 | XP_008760307.1 | 6 | 970 | |||
| XM_008762086.1 | XP_008760308.1 | 6 | 952 | |||
| XM_008762087.1 | XP_008760309.1 | 6 | 969 | |||
| GenBank mRNA | BC091215.1 | - | 5 | 718 | 7383613 | |

Mettl15
Genomic Map