siRNA Details
| Gene Symbol: Olr769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000371.1 | NP_001000371.1 | 1 | 334 | ||
| Gene Symbol: Olr775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000374.1 | NP_001000374.1 | 1 | 334 | ||

Olr769
Genomic Map