siRNA Details
| Gene Symbol: Emc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106495.1 | NP_001099965.1 | 3 | 415 | ||
| XM_006234704.2 | XP_006234766.1 | 4 | 1680 | |||
| XM_008762088.1 | XP_008760310.1 | 3 | 1676 | |||
| GenBank mRNA | FQ215850.1 | - | 3 | 355 | ||
| FQ234005.1 | - | 3 | 355 | |||

Emc4
Genomic Map