siRNA Details
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106497.1 | NP_001099967.1 | 2, 3 | 182 | ||
GenBank mRNA | BC166931.1 | - | 2, 3 | 138 | 6921277 | |
FQ210704.1 | - | 1, 2 | 82 | |||
FQ210780.1 | - | 1, 2 | 29 |
