siRNA Details
| Gene Symbol: Srp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106497.1 | NP_001099967.1 | 4 | 305 | ||
| GenBank mRNA | BC166931.1 | - | 4 | 261 | 6921277 | |
| FQ210704.1 | - | 3 | 205 | |||
| FQ210780.1 | - | 3 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp14| Gene Symbol: Srp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106497.1 | NP_001099967.1 | 4 | 305 | ||
| GenBank mRNA | BC166931.1 | - | 4 | 261 | 6921277 | |
| FQ210704.1 | - | 3 | 205 | |||
| FQ210780.1 | - | 3 | 152 | |||