siRNA Details
| Gene Symbol: Ino80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261404.1 | NP_001248333.1 | 11 | 1587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ino80| Gene Symbol: Ino80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261404.1 | NP_001248333.1 | 11 | 1587 | ||