siRNA Details
| Gene Symbol: ZBTB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143823.2 | NP_001137295.1 | 4 | 786 | ||
| NM_001243702.1 | NP_001230631.1 | 4 | 454 | |||
| NM_001243704.1 | NP_001230633.1 | 4 | 445 | |||
| NM_003409.4 | NP_003400.2 | 3 | 756 | |||
| GenBank mRNA | AK313236.1 | - | 3 | 599 | ||
| BC057282.1 | - | 4 | 632 | |||
| BC110519.2 | - | 2 | 340 | 40033453 | ||
| BI561601.1 | - | 4 | 459 | |||
| D89859.1 | - | 2 | 361 | |||
| DA817685.1 | - | 3 | 489 | |||
| Y12726.1 | - | 2 | 328 | |||

ZBTB14
Genomic Map