siRNA Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168527.1 | NP_001161999.1 | 9 | 1456 | ||
| XM_008762152.1 | XP_008760374.1 | 9 | 1425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Secisbp2l| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168527.1 | NP_001161999.1 | 9 | 1456 | ||
| XM_008762152.1 | XP_008760374.1 | 9 | 1425 | |||