siRNA Details
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168527.1 | NP_001161999.1 | 5 | 930 | ||
XM_008762152.1 | XP_008760374.1 | 5 | 887 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168527.1 | NP_001161999.1 | 5 | 930 | ||
XM_008762152.1 | XP_008760374.1 | 5 | 887 |