siRNA Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013919.1 | NP_001013941.1 | 2, 3 | 212 | ||
| XM_006234904.2 | XP_006234966.1 | 1, 2 | 667 | |||
| GenBank mRNA | BC083716.1 | - | 2, 3 | 212 | 7190694 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galk2| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013919.1 | NP_001013941.1 | 2, 3 | 212 | ||
| XM_006234904.2 | XP_006234966.1 | 1, 2 | 667 | |||
| GenBank mRNA | BC083716.1 | - | 2, 3 | 212 | 7190694 | |