siRNA Details
| Gene Symbol: Gpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168529.1 | NP_001162001.1 | 17 | 2050 | ||
| XM_006234956.2 | XP_006235018.1 | 20 | 2448 | |||
| XM_008762158.1 | XP_008760380.1 | 17 | 2792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpat2| Gene Symbol: Gpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168529.1 | NP_001162001.1 | 17 | 2050 | ||
| XM_006234956.2 | XP_006235018.1 | 20 | 2448 | |||
| XM_008762158.1 | XP_008760380.1 | 17 | 2792 | |||