siRNA Details
| Gene Symbol: Nphp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106506.1 | NP_001099976.1 | 14 | 2170 | ||
| XM_006234961.2 | XP_006235023.1 | 15 | 2414 | |||
| XM_006234963.2 | XP_006235025.1 | 15 | 2411 | |||
| XM_006234964.2 | XP_006235026.1 | 15 | 2411 | |||
| XM_006234965.2 | XP_006235027.1 | 15 | 2408 | |||
| GenBank mRNA | BC168839.1 | - | 10 | 1134 | ||

Nphp1
Genomic Map