siRNA Details
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106510.1 | NP_001099980.1 | 13 | 1887 |

More Information
Other Species Targeted

Gene Ontology Categories:

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106510.1 | NP_001099980.1 | 13 | 1887 |