siRNA Details
| Gene Symbol: Pced1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012348.1 | NP_001012348.1 | 4 | 707 | ||
| XM_006235015.2 | XP_006235077.1 | 6 | 1182 | |||
| XM_006235016.2 | XP_006235078.1 | 7 | 2554 | |||
| XM_006235017.2 | XP_006235079.1 | 6 | 1089 | |||
| GenBank mRNA | BC089908.1 | - | 4 | 707 | 7304556 | |

Pced1a
Genomic Map