siRNA Details
| Gene Symbol: Tmx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100529.1 | NP_001093999.1 | 2 | 361 | ||
| GenBank mRNA | CB582029.1 | - | 2 | 326 | ||
| CB760647.1 | - | 2 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmx4| Gene Symbol: Tmx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100529.1 | NP_001093999.1 | 2 | 361 | ||
| GenBank mRNA | CB582029.1 | - | 2 | 326 | ||
| CB760647.1 | - | 2 | 418 | |||