siRNA Details
| Gene Symbol: Sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106517.1 | NP_001099987.1 | 5 | 977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sptlc3| Gene Symbol: Sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106517.1 | NP_001099987.1 | 5 | 977 | ||