siRNA Details
| Gene Symbol: RGD1566320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106520.1 | NP_001099990.1 | 2 | 211 | ||
| XM_006235125.2 | XP_006235187.1 | 2 | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566320| Gene Symbol: RGD1566320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106520.1 | NP_001099990.1 | 2 | 211 | ||
| XM_006235125.2 | XP_006235187.1 | 2 | 372 | |||