siRNA Details
| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106523.1 | NP_001099993.1 | 3 | 470 | ||
| XM_006235213.2 | XP_006235275.1 | 4 | 1743 | |||
| XM_008762305.1 | XP_008760527.1 | 4 | 1743 | |||
| XM_008762306.1 | XP_008760528.1 | 4 | 1740 | |||

Cst7
Genomic Map