siRNA Details
Gene Symbol: Tspy26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106527.2 | NP_001099997.1 | 1 | 732 | ||
GenBank mRNA | BC168711.1 | - | 1 | 732 | 8366622 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspy26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106527.2 | NP_001099997.1 | 1 | 732 | ||
GenBank mRNA | BC168711.1 | - | 1 | 732 | 8366622 |