siRNA Details
| Gene Symbol: Raly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011958.1 | NP_001011958.1 | 3 | 335 | ||
| XM_008762314.1 | XP_008760536.1 | 3 | 725 | |||
| XM_008762315.1 | XP_008760537.1 | 2 | 443 | |||
| XM_008762316.1 | XP_008760538.1 | 3 | 403 | |||
| XM_008762317.1 | XP_008760539.1 | 2 | 439 | |||
| GenBank mRNA | BC087074.1 | - | 3 | 335 | 7134486 | |

Raly
Genomic Map