siRNA Details
| Gene Symbol: RGD1307752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013922.1 | NP_001013944.1 | 3 | 455 | ||
| GenBank mRNA | BC089919.1 | - | 3 | 455 | 7306699 | |
| FQ214713.1 | - | 3 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307752| Gene Symbol: RGD1307752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013922.1 | NP_001013944.1 | 3 | 455 | ||
| GenBank mRNA | BC089919.1 | - | 3 | 455 | 7306699 | |
| FQ214713.1 | - | 3 | 448 | |||